missing translation for 'onlineSavingsMsg'
Learn More

Chlortetracycline hydrochloride

Product Code. 15462929
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15462929 100 g 100g
15452929 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15462929 Supplier Thermo Scientific Alfa Aesar Supplier No. J60095.22
 Restricted Item
Estimated Shipment: 11-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Bacteriostatic antibiotic active against Gram-positive and Gram-negative bacteria | CAS: 64-72-2 | C22H24Cl2N2O8 | 515.34 g/mol

Chlortetracycline was used in fluorescence assays to detect the Ca+2 influx required for the completion of acrosome reaction in human spermatozoa.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlortetracycline was used in fluorescence assays to detect the Ca+2 influx required for the completion of acrosome reaction in human spermatozoa.

Solubility
Soluble in water

Notes
Light Sensitive. Store in dark. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-72-2
Molecular Formula C22H24Cl2N2O8
Molecular Weight (g/mol) 515.34
MDL Number MFCD00082440
InChI Key QYAPHLRPFNSDNH-CIVPRPTRSA-N
Synonym chlortetracycline hydrochloride, 4-epi-chlortetracycline hydrochloride, 2-amino hydroxy methylidene-7-chloro-4-dimethylamino-6,10,11,12a-tetrahydroxy-6-methyl-4,4a,5,5a-tetrahydrotetracene-1,3,12-trione hydrochloride, 2-azanyl oxidanyl methylidene-7-chloranyl-4-dimethylamino-6-methyl-6,10,11,12a-tetrakis oxidanyl-4,4a,5,5a-tetrahydrotetracene-1,3,12-trione hydrochloride
PubChem CID 66577600
IUPAC Name (4S,4aR,5aS,6S,12aR)-7-chloro-4-(dimethylamino)-1,6,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
SMILES CC1(C2CC3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C(C=CC(=C41)Cl)O)O)O)O)C(=O)N)N(C)C)O.Cl

Specifications

Melting Point 210°C to 215°C (decomposition)
Color Yellow to Brown
pH 2.3 to 3.3
Quantity 100 g
Beilstein 3858364
Sensitivity Light sensitive
Merck Index 14,2192
Solubility Information Soluble in water
Formula Weight 515.34
Physical Form Powder
Chemical Name or Material Chlortetracycline hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.