missing translation for 'onlineSavingsMsg'
Learn More

Chlortetracycline Hydrochloride, LoGiCal

Product Code. 30459180
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30459180 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30459180 Supplier LoGiCal Supplier No. LGCFOR0607.00
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chlortetracycline Hydrochloride
CAS 64-72-2
Analyte or Component Names Chlortetracycline Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C22 H23 Cl N2 O8 . Cl H
Certifications/Compliance ISO 9001
Synonym Chlortetracycline Hydrochloride, (4S,4aS,5aS,6S,12aS)-7-Chloro-4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-2-naphthacenecarboxamide Monohydrochloride, [4S-(4alpha,4aalpha,5aalpha,6beta,12aalpha)]-7-Chloro-4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-2-naphthacenecarboxamide Monohydrochloride, 7-Chlorotetracycline hydrochloride, 7-Chlorotetracycline monohydrochloride, Aureociclina, Aureocycline, Aureomycin hydrochloride, Aureomycin monohydrochloride, Aureovit 12C80, Aurofac 100, Biomycin hydrochloride, Chlortetracycline hydrochloride, Chlortetracyclinium chloride, Fermycin Soluble, Isphamycin, NSC 13252
InChI Formula InChI=1S/C22H23ClN2O8.ClH/c1-21(32)7-6-8-15(25(2)3)17(28)13(20(24)31)19(30)22(8,33)18(29)11(7)16(27)12-10(26)5-4-9(23)14(12)21;/h4-5,7-8,15,26,28-29,32-33H,6H2,1-3H3,(H2,24,31);1H/t7-,8-,15-,21-,22-;/m0./s1
Solution Type Neat
SMILES Cl.CN(C)[C@H]1[C@@H]2C[C@H]3C(=C(O)[C@]2(O)C(=O)C(=C1O)C(=O)N)C(=O)c4c(O)ccc(Cl)c4[C@@]3(C)O
IUPAC Name (4S,4aS,5aS,6S,12aS)-7-chloro-4-(dimethylamino)-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
Industry Type Clinical Testing
Molecular Weight (g/mol) 515.34
Quantity 10 mg
Formula Weight 514.091 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.