missing translation for 'onlineSavingsMsg'
Learn More

Chlortetracycline-13C-d3 Hydrochloride (Technical Grade), TRC

Product Code. 30357701
Change view
Click to view available options
Quantity:
0.5 mg
2.5 mg
5 mg
Unit Size:
0.5mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357701 0.5 mg 0.5mg
30362833 5 mg 5mg
30364824 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30357701 Supplier Toronto Research Chemicals Supplier No. TRCC4265040.5MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chlortetracycline-13C-d3 Hydrochloride (Technical Grade)
Molecular Formula C21 13C H20 D3 Cl N2 O8 . H Cl
Quantity 0.5 mg
Synonym (4S,4aS,5aS,6S,12aS)-7-Chloro-4-(dimethylamino-d6)-1,4,4a,5,5a,6,11,12a-octahydro-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-2-naphthacenecarboxamide-13C-d3 Hydrochloride, 7-Chlorotetracycline-13C-d3 Hydrochloride, 7-Chlorotetracycline-13C-d3 Monohydrochloride, Aureomycin-13C-d3 Hydrochloride, Biomycin-13C-d3 Hydrochloride, Chlortetracycline-13C-d3 Hydrochloride, Fermycin-13C-d3 Soluble, Isphamycin-13C-d3
InChI Formula InChI=1S/C22H23ClN2O8.ClH/c1-21(32)7-6-8-15(25(2)3)17(28)13(20(24)31)19(30)22(8,33)18(29)11(7)16(27)12-10(26)5-4-9(23)14(12)21;/h4-5,7-8,15,26,28-29,32-33H,6H2,1-3H3,(H2,24,31);1H/t7?,8-,15-,21-,22-;/m0./s1/i2+1D3;
SMILES OC1=CC=C(C2=C1C(C3=C([C@@](O)([C@]4(CC3[C@@]2(O)C)[H])C(C(C(N)=O)=C([C@H]4N([13C]([2H])([2H])[2H])C)O)=O)O)=O)Cl.Cl
IUPAC Name (4S,4aS,6S,12aS)-7-chloro-3,6,10,12,12a-pentahydroxy-6-methyl-4-(methyl(methyl-13C-d3)amino)-1,11-dioxo-1,4,4a,5,5a,6,11,12a-octahydrotetracene-2-carboxamide hydrochloride
Molecular Weight (g/mol) 519.35
Formula Weight 518.11
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.