missing translation for 'onlineSavingsMsg'
Learn More

Chlorpyrifos, TRC

Product Code. 18063310
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18063310 50 mg 50mg
18041120 250 mg 250mg
18044010 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18063310 Supplier Toronto Research Chemicals Supplier No. TRCC42530050MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2921-88-2
Molecular Formula C9 H11 Cl3 N O3 P S
Molecular Weight (g/mol) 350.59
Synonym Phosphorothioic acid, O,O-diethyl O-(3,5,6-trichloro-2-pyridyl) ester (7CI,8CI), Bonidel, CPF Zamba, Chlora, Chlora (insecticide), Chloroban, Chlorofet, Chloropyrifos, Chloropyrifos-ethyl, Chloropyriphos, Chlorpyrifos, Chlorpyrifos E, Chlorpyrifos-ethyl, Chlorpyriphos, Clorpiran, Clorpirifos, Coroban, Cyfos, Danusban, Dhanusban, Dowco 179, Durmet, Dursban, Dursban 10CR, Dursban 20 EC, Dursban 48EC, Dursban 4E, Dursban 75WG, Dursban L.O., Dursban Pro, Dursban R, Dursban TC, Dursband, Dursband 48, EF 1315, ENT 27311, Emperor, Empire 20, Equity, Ethyl chlorpyriphos, FE, Flower Power, GF 2153, Geodinfos, Gigant, Grofo, Grub Guard, Hilban, Killmaster, Klorpan, Klorpan 480 EC, LAF 1, Lentrek, Lock-On, Lorsban, Lorsban 15G, Lorsban 48E, Lorsban 50SL, Lorsban Advanced, Nufos, Nufos 4E, O,O-Diethyl O-(3,5,6-trichloro-2-pyridinyl)phosphorothioate, O,O-Diethyl O-(3,5,6-trichloro-2-pyridyl) phosphorothioate, O,O-Diethyl O-(3,5,6-trichloro-2-pyridyl) thiophosphate, O,O-Diethyl-O-3,5,6-trichloro-2-pyridylphosphorothionate, Pestban, Pirfos Glex, Pyriban, Pyrical, Pyrifos, Pyrinex, Pyrinex 480, Pyrinex 480EC, Radar, Radar (fungicide), Sabre, Saurus, Spannit, Spinter, Stipend, Tafaban, Terial, Terial 40L, Termicot, Termifos, Warhawk, XRM 429, XRM 5160, Xinnongba, suSCon, suSCon Blue, suSCon Plus, suScon Green
IUPAC Name diethoxy-sulfanylidene-(3,5,6-trichloropyridin-2-yl)oxy-λ^{5}-phosphane
SMILES CCOP(=S)(OCC)Oc1nc(Cl)c(Cl)cc1Cl

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C9H11Cl3NO3PS/c1-3-14-17(18,15-4-2)16-9-7(11)5-6(10)8(12)13-9/h5H,3-4H2,1-2H3
Formula Weight 348.9263
Percent Purity >95%
Chemical Name or Material Chlorpyrifos
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.