missing translation for 'onlineSavingsMsg'
Learn More

Chlorprothixene, MedChemExpress

Product Code. 16421406
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16421406

Brand: Medchem Express HYB0274

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Chlorprothixene is a dopamine and histamine receptors antagonist with Kis of 18 nM, 2.96 nM, 4.56 nM, 9 nM and 3.75 nM for hD1, hD2, hD3, hD5 and hH1 receptors, respectively. Antipsychotic activity.

Chlorprothixene is a dopamine and histamine receptors antagonist with Kis of 18 nM, 2.96 nM, 4.56 nM, 9 nM and 3.75 nM for hD1, hD2, hD3, hD5 and hH1 receptors, respectively. Antipsychotic activity.

Specifications

Chemical Name or Material Chlorprothixene
Physical Form Solid
CAS 113-59-7
Color Earth Yellow
Quantity 200 mg
Purity Grade Notes Research
Molecular Formula C18H18ClNS
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : 33.33 mg/mL (105.52 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES CN(C)CC/C=C1C2=C(SC3=C\1C=CC=C3)C=CC(Cl)=C2
Molecular Weight (g/mol) 315.86
Formula Weight 315.86
Percent Purity 99.13%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.