missing translation for 'onlineSavingsMsg'
Learn More

Chlorprothixene EP Impurity E, Mikromol™

Product Code. 17902566
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17902566 25 mg 25mg
17914336 100 mg 100mg
30462358 4 x 25 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17902566 Supplier Mikromol Supplier No. MM1321.050025
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Chlorprothixene EP Impurity E
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Chloro-9H-thioxanthen-9-one
CAS 86-39-5
Analyte or Component Names 2-Chloro-9H-thioxanthen-9-one
Shipping Condition Room Temperature
Molecular Formula C13 H7 Cl O S
Certifications/Compliance ISO 9001
Synonym Chlorprothixene Hydrochloride Imp. E (EP), Chlorprothixene Imp. E (EP), 2-Chloro-9H-thioxanthen-9-one, Chlorprothixene Hydrochloride Impurity E, Zuclopenthixol Decanoate Impurity B, Chlorprothixene Impurity E
InChI Formula InChI=1S/C13H7ClOS/c14-8-5-6-12-10(7-8)13(15)9-3-1-2-4-11(9)16-12/h1-7H
SMILES Clc1ccc2Sc3ccccc3C(=O)c2c1
IUPAC Name 2-chlorothioxanthen-9-one
Molecular Weight (g/mol) 246.71
Quantity 25 mg
Formula Weight 245.9906 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.