missing translation for 'onlineSavingsMsg'
Learn More

Chlorpromazine.HCl, 1mg/ml in Methanol (as free base), Lipomed

Product Code. 30458395
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30458395 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30458395 Supplier Lipomed Supplier No. LPMCPZ1752HC1LM
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Chlorpromazine.HCl, 1mg/ml in Methanol (as free base)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chlorpromazine Hydrochloride
CAS 69-09-0
Analyte or Component Names Chlorpromazine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C17 H19 Cl N2 S . Cl H
Concentration 1 mg/mL
Synonym Chlorpromazine Hydrochloride, 10 H-Phenothiazine-10-propanamine, 2-chloro-N,N-dimethyl-, monohydrochloride (9 CI), Phenothiazine, 2-chloro-10-[3-(dimethylamino)propyl]-, monohydrochloride (8 CI), 10-(3-Dimethylaminopropyl)-2-chlorophenothiazine monohydrochloride, 2-Chloro-10-(3-dimethylaminopropyl)phenothiazine monohydrochloride, Aminazin hydrochloride, Aminazin monohydrochloride, Ampliactil monohydrochloride, CPZ, Chloractil, Chlorazin, Chloropromazine hydrochloride, Chloropromazine monohydrochloride, Chlorpromazine monohydrochloride, Chlorpromazinium chloride, Hebanil, Hibanil, Hibernal, Klorproman, Klorpromex, Largactil, Largactil monohydrochloride, Largaktyl, Marazine, Megatil, Neurazine, Norcozine, Plegomazin, Promacid, Promapar, Propaphen, Propaphenin hydrochloride, Sonazine, Taroctyl, Thorazine hydrochloride, Torazina, Tranzene
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES Cl.CN(C)CCCN1c2ccccc2Sc3ccc(Cl)cc13
IUPAC Name 3-(2-chlorophenothiazin-10-yl)-N,N-dimethylpropan-1-amine;hydrochloride
Solvent or Matrix Methanol
Industry Type Pharmaceutical
Molecular Weight (g/mol) 355.33
Quantity 1 mL
Formula Weight 354.0724 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.