missing translation for 'onlineSavingsMsg'
Learn More

Chlorpheniramine Maleate Salt, TRC

Product Code. 30356781
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30356781 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30356781 Supplier Toronto Research Chemicals Supplier No. TRCC424300500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chlorphenamine Maleate
CAS 113-92-8
Purity Grade Notes HPLC
Molecular Formula C16 H19 Cl N2 . C4 H4 O4
Quantity 500 mg
Synonym 2-Pyridinepropanamine, γ-(4-chlorophenyl)-N,N-dimethyl-, (Z)-2-butenedioate (1:1), Pyridine, 2-[p-chloro-α-[2-(dimethylamino)ethyl]benzyl]-, maleate (1:1) (8CI), (±)-Chlorpheniramine maleate, 1-Parachlorophenyl-1-(2-pyridyl)-3-dimethylaminopropane maleate, 1-p-Chlorophenyl-1-(2-pyridyl)-3-dimethylaminopropane maleate, Allergin, Allergisan, Anallerge 4, Antagonate, C-Meton, Calimal, Carbinoxamide maleate, Chlor-Trimeton, Chlor-Tripolon, Chlorophenamine, Chloroprophenpyridamine maleate, Chlorphenamine hydrogen maleate, Chlorphenamine maleate, Chlorpheniramine maleate, Chlorprophenpyridamine maleate, Cloropiril, Histadur, Histadur dura-tabs, Histamil, Histaspan, Ibioton, Iramine, Lorphen, Neorestamin, Niramine, Pirafene, Piriex, Piriton, Puermin, Pyridamal 100, Synistamin, Teldrin, dl-Chlorpheniramine maleate
InChI Formula InChI=1S/C16H19ClN2.C4H4O4/c1-19(2)12-10-15(16-5-3-4-11-18-16)13-6-8-14(17)9-7-13;5-3(6)1-2-4(7)8/h3-9,11,15H,10,12H2,1-2H3;1-2H,(H,5,6)(H,7,8)/b;2-1-
SMILES CN(C)CCC(c1ccc(Cl)cc1)c2ccccn2.OC(=O)\C=C/C(=O)O
IUPAC Name (Z)-but-2-enedioic acid;3-(4-chlorophenyl)-N,N-dimethyl-3-pyridin-2-ylpropan-1-amine
Molecular Weight (g/mol) 390.86
Formula Weight 390.1346
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.