missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorpheniramine Maleate Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Chlorphenamine Maleate |
| CAS | 113-92-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C16 H19 Cl N2 . C4 H4 O4 |
| Quantity | 250 mg |
| Synonym | 2-Pyridinepropanamine, γ-(4-chlorophenyl)-N,N-dimethyl-, (Z)-2-butenedioate (1:1), Pyridine, 2-[p-chloro-α-[2-(dimethylamino)ethyl]benzyl]-, maleate (1:1) (8CI), (±)-Chlorpheniramine maleate, 1-Parachlorophenyl-1-(2-pyridyl)-3-dimethylaminopropane maleate, 1-p-Chlorophenyl-1-(2-pyridyl)-3-dimethylaminopropane maleate, Allergin, Allergisan, Anallerge 4, Antagonate, C-Meton, Calimal, Carbinoxamide maleate, Chlor-Trimeton, Chlor-Tripolon, Chlorophenamine, Chloroprophenpyridamine maleate, Chlorphenamine hydrogen maleate, Chlorphenamine maleate, Chlorpheniramine maleate, Chlorprophenpyridamine maleate, Cloropiril, Histadur, Histadur dura-tabs, Histamil, Histaspan, Ibioton, Iramine, Lorphen, Neorestamin, Niramine, Pirafene, Piriex, Piriton, Puermin, Pyridamal 100, Synistamin, Teldrin, dl-Chlorpheniramine maleate |
| InChI Formula | InChI=1S/C16H19ClN2.C4H4O4/c1-19(2)12-10-15(16-5-3-4-11-18-16)13-6-8-14(17)9-7-13;5-3(6)1-2-4(7)8/h3-9,11,15H,10,12H2,1-2H3;1-2H,(H,5,6)(H,7,8)/b;2-1- |
| SMILES | CN(C)CCC(c1ccc(Cl)cc1)c2ccccn2.OC(=O)\C=C/C(=O)O |
| IUPAC Name | (Z)-but-2-enedioic acid;3-(4-chlorophenyl)-N,N-dimethyl-3-pyridin-2-ylpropan-1-amine |
| Molecular Weight (g/mol) | 390.86 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?