missing translation for 'onlineSavingsMsg'
Learn More

Chlorpheniramine maleate, MedChemExpress

Product Code. p-200101119
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16482119

Brand: Medchem Express HYB0286A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Chlorpheniramine maleate is an histamine H1 receptor antagonist with IC50 of 12 nM.

Chlorpheniramine maleate is an histamine H1 receptor antagonist with IC50 of 12 nM.

Specifications

Chemical Name or Material Chlorpheniramine maleate
CAS 113-92-8
Color White
Purity Grade Notes Research
Molecular Formula C20H23ClN2O4
Quantity 1 g
Synonym Chlorphenamine maleate
Solubility Information DMSO : ≥ 100 mg/mL (255.85 mM) ∣H2O : 33.33 mg/mL (85.27 mM; Need ultrasonic)
SMILES CN(C)CCC(C1=CC=C(Cl)C=C1)C2=CC=CC=N2.O=C(O)/C=C\C(O)=O
Molecular Weight (g/mol) 390.86
Formula Weight 390.86
Percent Purity 99.91%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.