missing translation for 'onlineSavingsMsg'
Learn More

Chlorpheniramine maleate, MedChemExpress

Product Code. 16482119
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16482119 1 g 1g
16470858 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16482119 Supplier Medchem Express Supplier No. HYB0286A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Chlorpheniramine maleate is an histamine H1 receptor antagonist with IC50 of 12 nM.

Chlorpheniramine maleate is an histamine H1 receptor antagonist with IC50 of 12 nM.

Specifications

Chemical Name or Material Chlorpheniramine maleate
CAS 113-92-8
Color White
Purity Grade Notes Research
Molecular Formula C20H23ClN2O4
Quantity 1 g
Synonym Chlorphenamine maleate
Solubility Information DMSO : ≥ 100 mg/mL (255.85 mM) ∣H2O : 33.33 mg/mL (85.27 mM; Need ultrasonic)
SMILES CN(C)CCC(C1=CC=C(Cl)C=C1)C2=CC=CC=N2.O=C(O)/C=C\C(O)=O
Molecular Weight (g/mol) 390.86
Formula Weight 390.86
Percent Purity 99.91%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.