missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorphenesin Carbamate, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Chlorphenesin Carbamate |
| CAS | 886-74-8 |
| Analyte or Component Names | Chlorphenesin Carbamate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C10 H12 Cl N O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Chlorphenesin Carbamate, 1,2-Propanediol, 3-(4-chlorophenoxy)-, 1-carbamate, 1,2-Propanediol, 3-(p-chlorophenoxy)-, 1-carbamate (8CI), Carbamic acid, 3-(p-chlorophenoxy)-2-hydroxypropyl ester (7CI), 3-(4-Chlorophenoxy)-2-hydroxypropyl carbamate, 3-(p-Chlorophenoxy)-2-hydroxypropyl carbamate, 3-[p-Chlorophenoxy]-1,2-propanediol-1-carbamate, Chlorphenesin carbamate, Maolate, NSC 82943, Rhnesicn, Rinlaxer, U 19646, U-19,646 |
| InChI Formula | InChI=1S/C10H12ClNO4/c11-7-1-3-9(4-2-7)15-5-8(13)6-16-10(12)14/h1-4,8,13H,5-6H2,(H2,12,14) |
| Solution Type | Neat |
| SMILES | NC(=O)OCC(O)COc1ccc(Cl)cc1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction