missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorphenesin 2-Carbamate (3-(4-Chlorophenoxy)-1,2-propanediol 2-Carbamate), Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Chlorphenesin 2-Carbamate (3-(4-Chlorophenoxy)-1,2-propanediol 2-Carbamate)
Specifications
Specifications
| Chemical Name or Material | Chlorphenesin 2-Carbamate |
| CAS | 61514-96-3 |
| Analyte or Component Names | Chlorphenesin 2-Carbamate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C10 H12 Cl N O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 1,2-Propanediol, 3-(4-chlorophenoxy)-, 2-carbamate (9CI), Carbamic acid, 2-(p-chlorophenoxy)-1-(hydroxymethyl)ethyl ester (7CI), Chlorphenesin 2-Carbamate |
| InChI Formula | InChI=1S/C10H12ClNO4/c11-7-1-3-8(4-2-7)15-6-9(5-13)16-10(12)14/h1-4,9,13H,5-6H2,(H2,12,14) |
| SMILES | NC(=O)OC(CO)COc1ccc(Cl)cc1 |
| IUPAC Name | [1-(4-chlorophenoxy)-3-hydroxypropan-2-yl] carbamate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction