missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorphenamine Maleate, Mikromol™
Description
Chlorphenamine Maleate
Specifications
Specifications
| Chemical Name or Material | Chlorphenamine Maleate |
| CAS | 113-92-8 |
| Analyte or Component Names | Chlorphenamine Maleate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H19 Cl N2 . C4 H4 O4 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Brompheniramine Maleate Imp. B (EP), Brompheniramine Imp. B (EP), Chlorphenamine Maleate, Brompheniramine Maleate Imp. A (EP) as Maleate, Chlorpheniramine Maleate, gamma-(4-Chlorophenyl)-N,N-dimethyl-2-pyridinepropanamine (2Z)-2-butenedioate (1:1), 2-[p-Chloro-alpha-[2-(dimethylamino)ethyl]benzyl]pyridine maleate (1:1), (+/-)-Chlorpheniramine maleate, 1-Parachlorophenyl-1-(2-pyridyl)-3-dimethylaminopropane maleate, 1-p-Chlorophenyl-1-(2-pyridyl)-3-dimethylaminopropane maleate, Allergin, Allergisan, Antagonate, C-Meton, Carbinoxamide maleate, Chlor-Trimeton, Chlor-Tripolon, Chlorophenamine, Chloroprophenpyridamine maleate, Chlorphenamine hydrogen maleate, Chlorphenamine maleate, Chlorpheniramine maleate, Chlorprophenpyridamine maleate, Cloropiril, Histadur, Histadur dura-tabs, Histaspan, Ibioton, Lorphen, Neorestamin, Piriex, Piriton, Puermin, Pyridamal 100, Synistamin, Teldrin, dl-Chlorpheniramine maleate, Brompheniramine Maleate Impurity A as Maleate, Brompheniramine Impurity A as Maleate |
| InChI Formula | InChI=1S/C16H19ClN2.C4H4O4/c1-19(2)12-10-15(16-5-3-4-11-18-16)13-6-8-14(17)9-7-13;5-3(6)1-2-4(7)8/h3-9,11,15H,10,12H2,1-2H3;1-2H,(H,5,6)(H,7,8)/b;2-1- |
| SMILES | CN(C)CCC(c1ccc(Cl)cc1)c2ccccn2.OC(=O)\C=C/C(=O)O |
| IUPAC Name | (Z)-but-2-enedioic acid;3-(4-chlorophenyl)-N,N-dimethyl-3-pyridin-2-ylpropan-1-amine |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?