missing translation for 'onlineSavingsMsg'
Learn More

Chlorozol Black E (Technical Grade), TRC

Product Code. 30361830
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
Unit Size:
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30361830 25 g 25g
30349276 50 g 50g
30353123 100 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30361830 Supplier Toronto Research Chemicals Supplier No. TRCC42796525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1937-37-7
Molecular Formula C34 H25 N9 O7 S2 . 2 Na
Molecular Weight (g/mol) 781.73
Synonym 2,7-Naphthalenedisulfonic acid, 4-amino-3-[2-[4'-[2-(2,4-diaminophenyl)diazenyl][1,1'-biphenyl]-4-yl]diazenyl]-5-hydroxy-6-(2-phenyldiazenyl)-, sodium salt (1:2) (ACI), 2,7-Naphthalenedisulfonic acid, 4-amino-3-[[4'-[(2,4-diaminophenyl)azo][1,1'-biphenyl]-4-yl]azo]-5-hydroxy-6-(phenylazo)-, disodium salt (9CI), C.I. Direct Black 38 (7CI), C.I. Direct Black 38, disodium salt (8CI), Chlorazol Black E (6CI), Ahco Direct Black GX, Airedale Black ED, Aizen Direct Deep Black EH, Aizen Direct Deep Black GH, Aizen Direct Deep Black RH, Amanil Black GL, Amanil Black WD, Apomine Black GX, Atlantic Black BD, Atlantic Black C, Atlantic Black E, Atlantic Black EA, Atlantic Black GAC, Atlantic Black GG, Atlantic Black GXCW, Atlantic Black GXOO, Atlantic Black SD, Atul Direct Black E, Azine Deep Black EW, Azocard Black EW, Azomine Black EWO, Belamine Black GX, Bencidal Black E, Benzanil Black E, Benzo Deep Black E, Benzo Leather Black E, Benzoform Black BCN-CF, Black 2EMBL, Black 4EMBL, Black FR, Brasilamina Black GN, Brilliant Chrome Leather Black H, C.I. 30235, Calcomine Black, Calcomine Black EXL, Carbide Black E, Chloramine Black C, Chloramine Black EC, Chloramine Black ERT, Chloramine Black EX, Chloramine Black EXR, Chloramine Black XO, Chloramine Carbon Black S, Chloramine Carbon Black SJ, Chloramine Carbon Black SN, Chlorazol Black EA, Chlorazol Black EN, Chlorazol Burl Black E, Chlorazol Leather Black ENP, Chlorazol Silk Black G, Chlorozol Black E, Chrome Leather Black E, Chrome Leather Black EC, Chrome Leather Black EM, Chrome Leather Black G, Chrome Leather Brilliant Black ER, Coir Deep Black C, Columbia Black EP, Coranil Direct Black F, Diacotton Deep Black, Diacotton Deep Black RX, Diamine Deep Black EC, Diamine Direct Black E, Diaphtamine Black V, Diazine Black E, Diazine Direct Black E, Diazine Direct Black G, Diazol Black 2V, Diphenyl Deep Black G, Direct Black 38, Direct Black A, Direct Black BN, Direct Black BRN, Direct Black CX, Direct Black CXR, Direct Black E, Direct Black EW, Direct Black EX, Direct Black FR, Direct Black GAC, Direct Black GW, Direct Black GX, Direct Black GXR, Direct Black JET, Direct Black Meta, Direct Black methyl, Direct Black N, Direct Black RX, Direct Black SD, Direct Black WS, Direct Black Z, Direct Black ZSh, Direct Deep Black E, Direct Deep Black E Extra, Direct Deep Black E-EX, Direct Deep Black EA-CF, Direct Deep Black EAC, Direct Deep Black EW, Direct Deep Black EX, Direct Deep Black WX, Enianil Black CN, Erie Black B, Erie Black BF, Erie Black GAC, Erie Black GXOO, Erie Black JET, Erie Black NUG, Erie Black RXOO, Erie Brilliant Black S, Erie Fibre Black VP, Fenamin Black E, Fibre Black VF, Fixanol Black E, Formaline Black C, Formic Black C, Formic Black CW, Formic Black EA, Formic Black MTG, Formic Black TG, Hispamin Black EF, Interchem Direct Black Z, Kayaku Direct Deep Black EX, Kayaku Direct Deep Black GX, Kayaku Direct Deep Black S, Kayaku Direct Leather Black EX, Kayaku Direct Special Black AAX, Lurazol Black BA, Meta Black, Mitsui Direct Black EX, Mitsui Direct Black GX, Nippon Deep Black, Nippon Deep Black GX, NSC 8679, Paper Black BA, Paper Black T, Paper Deep Black C, Paramine Black B, Paramine Black E, Peeramine Black E, Peeramine Black GXOO, Phenamine Black BCN-CF, Phenamine Black CL, Phenamine Black E, Phenamine Black E 200, Pheno Black EP, Pheno Black SGN, Pontamine Black E, Pontamine Black EBN, Sandopel Black EX, Seristan Black B, Telon Fast Black E, Tertrodirect Black E, Tertrodirect Black EFD, Tetrazo Deep Black G, Union Black EM, Vondacel Black N
IUPAC Name disodium;4-amino-3-[(E)-[4-[4-[(E)-(2,4-diaminophenyl)diazenyl]phenyl]phenyl]diazenyl]-5-hydroxy-6-[(E)-phenyldiazenyl]naphthalene-2,7-disulfonate
SMILES [Na+].[Na+].Nc1ccc(N=Nc2ccc(cc2)c3ccc(cc3)N=Nc4c(N)c5c(O)c(N=Nc6ccccc6)c(cc5cc4S(=O)(=O)[O-])S(=O)(=O)[O-])c(N)c1

Specifications

Quantity 25 g
InChI Formula InChI=1S/C34H27N9O7S2.2Na/c35-22-10-15-27(26(36)18-22)41-38-24-11-6-19(7-12-24)20-8-13-25(14-9-20)40-42-32-28(51(45,46)47)16-21-17-29(52(48,49)50)33(34(44)30(21)31(32)37)43-39-23-4-2-1-3-5-23;;/h1-18,44H,35-37H2,(H,45,46,47)(H,48,49,50);;/q;2*+1/p-2
Formula Weight 781.1114
Chemical Name or Material Direct Black 38
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.