missing translation for 'onlineSavingsMsg'
Learn More

Chloroxylenol, TRC

Product Code. 30357078
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30357078 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30357078 Supplier Toronto Research Chemicals Supplier No. TRCC42795050G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chloroxylenol
CAS 88-04-0
Molecular Formula C8 H9 Cl O
Quantity 50 g
Synonym 3,5-Xylenol, 4-chloro- (6CI,8CI), 4-Chloro-3,5-dimethylphenol, 2,6-Dimethyl-4-hydroxy-1-chlorobenzene, 2-Chloro-5-hydroxy-1,3-dimethylbenzene, 2-Chloro-5-hydroxy-m-xylene, 3,5-Dimethyl-p-chlorophenol, 4-Chloro-3,5-Xylenol, 4-Chloro-5,3-dimethylphenol, 4-Chloro-m-xylenol, Benzytol, Camel, Camel (pesticide), Chloroxylenol, Desson, Dettol, Dettol (biocide), Ecotru, Espadol, Finecide C 2000, Hokubarine E 400, Husept Extra, NSC 4971, Nipacide PX, Nipacide PX-R, Ottasept, Ottasept Extra, PCMX, Parametaxylenol, RBA 777, p-Chloro-3,5-xylenol, p-Chloro-m-Xylenol
InChI Formula InChI=1S/C8H9ClO/c1-5-3-7(10)4-6(2)8(5)9/h3-4,10H,1-2H3
SMILES Cc1cc(O)cc(C)c1Cl
IUPAC Name 4-chloro-3,5-dimethylphenol
Molecular Weight (g/mol) 156.61
Formula Weight 156.0342
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.