missing translation for 'onlineSavingsMsg'
Learn More

Chloroxylenol, MedChemExpress

Product Code. 16491108
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16491108 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16491108 Supplier Medchem Express Supplier No. HYB1414
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Chloroxylenol is a broad spectrum antimicrobial chemical compound used to control bacteria, algae, fungi and virus.

Chloroxylenol is a broad spectrum antimicrobial chemical compound used to control bacteria, algae, fungi and virus.

Specifications

Chemical Name or Material Chloroxylenol
Physical Form Solid
CAS 88-04-0
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C8H9ClO
For Use With (Application) COVID-19-anti-virus
Synonym 4-Chloro-3,5-dimethylphenol PCMX
Solubility Information DMSO : ≥ 100 mg/mL (638.53 mM)
SMILES OC1=CC(C)=C(Cl)C(C)=C1
Molecular Weight (g/mol) 156.61
Formula Weight 156.61
Percent Purity 99.12%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.