missing translation for 'onlineSavingsMsg'
Learn More

Chloroxine, TRC

Product Code. 30004061
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004061 10 g 10g
17920785 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30004061 Supplier Toronto Research Chemicals Supplier No. TRCC42413510G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 773-76-2
Molecular Formula C9 H5 Cl2 N O
Molecular Weight (g/mol) 214.05
Synonym 5,7-Dichloroquinolin-8-ol, Clioquinol Imp. B (EP)
IUPAC Name 5,7-dichloroquinolin-8-ol
SMILES Oc1c(Cl)cc(Cl)c2cccnc12

Specifications

Quantity 10 g
InChI Formula InChI=1S/C9H5Cl2NO/c10-6-4-7(11)9(13)8-5(6)2-1-3-12-8/h1-4,13H
Formula Weight 212.9748
Percent Purity >95%
Chemical Name or Material 5,7-Dichloroquinolin-8-ol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.