missing translation for 'onlineSavingsMsg'
Learn More

Chloroxine, MedChemExpress

Product Code. 16428226
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16428226 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16428226 Supplier Medchem Express Supplier No. HYB0295
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Chloroxine is one of the important 8-hydroxyquinoline derivative. Chloroxine has effective antibacterial, antifungal, antiprotozoal and antiamoebic activities, especially used in treating the intestinal amebiasis. Chloroxine is also used in the treatment of dandruff and seborrheic dermatitis of the scalp.

Chloroxine is one of the important 8-hydroxyquinoline derivative. Chloroxine has effective antibacterial, antifungal, antiprotozoal and antiamoebic activities, especially used in treating the intestinal amebiasis. Chloroxine is also used in the treatment of dandruff and seborrheic dermatitis of the scalp.

Specifications

Chemical Name or Material Chloroxine
Physical Form Solid
CAS 773-76-2
Color Taupe
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C9H5Cl2NO
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 33.33 mg/mL (155.71 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES OC1=C2N=CC=CC2=C(Cl)C=C1Cl
Molecular Weight (g/mol) 214.05
Formula Weight 214.05
Percent Purity 99.38%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.