missing translation for 'onlineSavingsMsg'
Learn More

Chlorotris(triphenylphosphine)cobalt(I), 97%

Product Code. 15451618
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15451618 25 g 25g
15431618 1 g 1g
15441618 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15451618 Supplier Thermo Scientific Alfa Aesar Supplier No. H55395.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 26305-75-9 | C54H45ClCoP3 | 881.26 g/mol

A stoichiometric reducing agent that is used in the radical dimerization of halogenated organic molecules.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A stoichiometric reducing agent that is used in the radical dimerization of halogenated organic molecules.

Solubility
Insoluble in water.

Notes
Air & Moisture Sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26305-75-9
Molecular Formula C54H45ClCoP3
Molecular Weight (g/mol) 881.26
MDL Number MFCD00015864
InChI Key RHNBOEDTKLKUEB-UHFFFAOYSA-M
Synonym chlorotris triphenylphosphine cobalt i, cobalt i tris triphenylphosphine chloride
PubChem CID 71431130
IUPAC Name cobalt;triphenylphosphane;chloride
SMILES [Cl-].[Co+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 135°C to 139°C
Quantity 25 g
Sensitivity Air and moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 881.26
Percent Purity 97%
Chemical Name or Material Chlorotris(triphenylphosphine)cobalt(I)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.