missing translation for 'onlineSavingsMsg'
Learn More

Chlorotriphenylstannane, TRC

Product Code. 30361973
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30361973 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30361973 Supplier Toronto Research Chemicals Supplier No. TRCC422250100MG
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fentin-chloride (Triphenyltin chloride)
CAS 639-58-7
Molecular Formula C18 H15 Cl Sn
Quantity 100 mg
Synonym Stannane, chlorotriphenyl-, Triphenyltin chloride (6CI), Chlorotriphenylstannane, Aquatin, Brestanol, Chlorotriphenyltin, Fentin chloride, Fentin-chloride, GC 8993, General Chemicals 8993, HOE 2872, LS 4442, NSC 1214, NSC 43675, TPTC, Triphenylchlorostannane, Triphenylchlorotin, Triphenylstannyl chloride, Triphenyltin monochloride, Wenqukexing
InChI Formula InChI=1S/3C6H5.ClH.Sn/c3*1-2-4-6-5-3-1;;/h3*1-5H;1H;/q;;;;+1/p-1
SMILES Cl[Sn](c1ccccc1)(c2ccccc2)c3ccccc3
IUPAC Name chloro(triphenyl)stannane
Molecular Weight (g/mol) 385.47
Formula Weight 385.9884
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.