missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorotriphenylmethane, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Chlorotriphenylmethane is used as laboratory chemical. It acts as a protecting reagent for thiols, alcohols and amines. It is used in organic synthesis for initiating trityl group protection and for preparing S-trityl-L-cysteine (STLC), a inhibitor of Eg5 prevents mitotic progression.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Chlorotriphenylmethane is used as laboratory chemical. It acts as a protecting reagent for thiols, alcohols and amines. It is used in organic synthesis for initiating trityl group protection and for preparing S-trityl-L-cysteine (STLC), a inhibitor of Eg5 prevents mitotic progression.
Solubility
Insoluble in water. Soluble in chloroform, hexane, benzene, ether, acetone and tetrahydrofuran.
Notes
Incompatible with strong oxidizing agents.
Chlorotriphenylmethane is used as laboratory chemical. It acts as a protecting reagent for thiols, alcohols and amines. It is used in organic synthesis for initiating trityl group protection and for preparing S-trityl-L-cysteine (STLC), a inhibitor of Eg5 prevents mitotic progression.
Solubility
Insoluble in water. Soluble in chloroform, hexane, benzene, ether, acetone and tetrahydrofuran.
Notes
Incompatible with strong oxidizing agents.
Specifications
Melting Point | 110°C to 114°C |
Boiling Point | 230°C to 235°C (20 mmHg) |
Linear Formula | (C6H5)3CCl |
Quantity | 25 g |
UN Number | UN3261 |
Beilstein | 397363 |
Sensitivity | Moisture sensitive |
Solubility Information | Insoluble in water. Soluble in chloroform,hexane,benzene,ether,acetone and tetrahydrofuran. |
Formula Weight | 278.78 |
Percent Purity | 98% |
Show More |
Safety and Handling
Signal Word
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 1
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'