missing translation for 'onlineSavingsMsg'
Learn More

Chlorotrimethylsilane, for Gas Chromatography, Fisher Chemical

Product Code. 10745731 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
250 g
Packaging:
Amber glass bottle
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10745731 250 g Amber glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10745731 Supplier Fisher Chemical Supplier No. T/3520/50

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 75-77-4
Melting Point -58°C
Color Colorless
Boiling Point 57°C
Packaging Amber glass bottle
Molecular Formula C3H9ClSi
MDL Number 502
Quantity 250 g
Synonym trimethylchlorosilane, trimethylsilyl chloride, silane, chlorotrimethyl, trimethyl chlorosilane, monochlorotrimethylsilicon, tmcs, chloro trimethyl silane, silane, trimethylchloro, silicane, chlorotrimethyl, tmscl
InChI Key IJOOHPMOJXWVHK-UHFFFAOYSA-N
SMILES C[Si](C)(C)Cl
IUPAC Name chloro(trimethyl)silane
Molecular Weight (g/mol) 108.64
PubChem CID 6397
ChEBI CHEBI:85069
Formula Weight 108.64g/mol
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Chlorotrimethylsilane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.