missing translation for 'onlineSavingsMsg'
Learn More

Chlorotrimethylsilane, 98%

Product Code. 11493900
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493900 25 mL 25mL
11400244 100 mL 100mL
11410244 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11493900 Supplier Thermo Scientific Alfa Aesar Supplier No. A13651.AC
Curiosity Goes Beyond
Use promo code 27966
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 75-77-4 | C3H9ClSi | 108.64 g/mol

Chlorotrimethylsilane is used to silanize laboratory glassware, which makes the surfaces more lipophilic. It is used in the production of trimethylsilyl halides, pseudohalides and various organic silicon compounds. It is also used to produce hexamethyldisilane by reduction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorotrimethylsilane is used to silanize laboratory glassware, which makes the surfaces more lipophilic. It is used in the production of trimethylsilyl halides, pseudohalides and various organic silicon compounds. It is also used to produce hexamethyldisilane by reduction.

Solubility
Miscible with ether, benzene, diethylether and perchloroethylene.

Notes
Incompatible with water, strong acids, strong oxidizing agents, strong bases, alcohols, aldehydes, amines, ketones and esters.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 75-77-4
Molecular Formula C3H9ClSi
Molecular Weight (g/mol) 108.64
MDL Number MFCD00000502
InChI Key IJOOHPMOJXWVHK-UHFFFAOYSA-N
Synonym trimethylchlorosilane, trimethylsilyl chloride, silane, chlorotrimethyl, trimethyl chlorosilane, monochlorotrimethylsilicon, tmcs, chloro trimethyl silane, silane, trimethylchloro, silicane, chlorotrimethyl, tmscl
PubChem CID 6397
ChEBI CHEBI:85069
IUPAC Name chloro(trimethyl)silane
SMILES C[Si](C)(C)Cl

Spezifikation

Boiling Point 56°C to 58°C
Melting Point -58°C
Quantity 25 mL
Linear Formula (CH3)3SiCl
UN Number UN1298
Beilstein 1209232
Flash Point −27°C (−16°F)
Solubility Information Miscible with ether,benzene,diethylether and perchloroethylene.
Formula Weight 108.64
Percent Purity ≥98%
Odor Chlorine-like
Refractive Index 1.389
Sensitivity Moisture sensitive
Density 0.856
Chemical Name or Material Chlorotrimethylsilane
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons compliance-icons
Produktidentifikator
  • Chlorotrimethylsilane
Signalwort
  • Danger
Gefahrenkategorie
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
Gefahrenhinweis
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • EUH014-Reacts violently with water.
  • EUH071-Corrosive to the respiratory tract.
Sicherheitshinweis
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.
Ergänzende Informationen
  • MIXTURE LIST-Contain: Trimethylchlorosilane

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.