missing translation for 'onlineSavingsMsg'
Learn More

Chlorotriisopropylsilane, 97+%

Product Code. 11493617
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11493617 50 g 50g
11483617 10 g 10g
11403627 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11493617 Supplier Thermo Scientific Alfa Aesar Supplier No. A17376.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13154-24-0 | C9H21ClSi | 192.80 g/mol

Chlorotriisopropylsilane is used as a silylating agent in neucleotide synthesis. It is used as an intermediate in the manufacture of chemical substances such as pharmaceuticals and in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorotriisopropylsilane is used as a silylating agent in neucleotide synthesis. It is used as an intermediate in the manufacture of chemical substances such as pharmaceuticals and in organic synthesis.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents, strong bases, alcohols and amines.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13154-24-0
Molecular Formula C9H21ClSi
Molecular Weight (g/mol) 192.80
MDL Number MFCD00009656
InChI Key KQIADDMXRMTWHZ-UHFFFAOYSA-N
Synonym triisopropylsilyl chloride, chlorotriisopropylsilane, triisopropylchlorosilane, chlorotris propan-2-yl silane, chloro triisopropyl silane, silane, chlorotris 1-methylethyl, tipscl, triisopropylsilylchloride, triisopropyl chlorosilane, triisopropylchlorosilane, redistilled
PubChem CID 139400
IUPAC Name chloro-tri(propan-2-yl)silane
SMILES CC(C)[Si](Cl)(C(C)C)C(C)C

Specifications

Boiling Point 200°C
Quantity 50 g
UN Number UN2987
Beilstein 1737446
Flash Point 62°C (144°F)
Solubility Information Miscible with water.
Formula Weight 192.81
Percent Purity ≥97%
Odor Acrid
Refractive Index 1.4535
Sensitivity Moisture sensitive
Density 0.908
Chemical Name or Material Chlorotriisopropylsilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.