missing translation for 'onlineSavingsMsg'
Learn More

Chlorotriisopropylsilane, 97%

Product Code. 10286940 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10286940 50 g Glass bottle 50g
10012960 10 g Glass bottle 10g
11995821 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10286940 Supplier Thermo Scientific Acros Supplier No. 221260500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13154-24-0 | C9H21ClSi | 192.80 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13154-24-0
Molecular Formula C9H21ClSi
Molecular Weight (g/mol) 192.80
MDL Number MFCD00009656
InChI Key KQIADDMXRMTWHZ-UHFFFAOYSA-N
Synonym triisopropylsilyl chloride, chlorotriisopropylsilane, triisopropylchlorosilane, chlorotris propan-2-yl silane, chloro triisopropyl silane, silane, chlorotris 1-methylethyl, tipscl, triisopropylsilylchloride, triisopropyl chlorosilane, triisopropylchlorosilane, redistilled
PubChem CID 139400
SMILES CC(C)[Si](Cl)(C(C)C)C(C)C

Specifications

Color Colorless to Yellow
Density 0.9020g/mL
Boiling Point 198.0°C
Flash Point 63°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula [(CH3)2CH]3SiCl
Refractive Index 1.4520 to 1.4540
Quantity 50 g
Specific Gravity 0.902
Solubility Information Solubility in water: reacts
IUPAC Name chlorotris(propan-2-yl)silane
Formula Weight 192.81
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Chlorotriisopropylsilane
Show More Show Less
compliance-icons
Product Identifier
  • Chlorotriisopropylsilane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.