missing translation for 'onlineSavingsMsg'
Learn More

Chlorotoluron, TRC

Product Code. 30137534
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137534 10 g 10g
17951575 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137534 Supplier Toronto Research Chemicals Supplier No. TRCC42160010G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15545-48-9
Molecular Formula C10 H13 Cl N2 O
Molecular Weight (g/mol) 212.68
Synonym Urea, 3-(3-chloro-p-tolyl)-1,1-dimethyl- (6CI,8CI), N'-(3-Chloro-4-methylphenyl)-N,N-dimethylurea, 1,1-Dimethyl-3-(3-chloro-4-methylphenyl)urea, 1-(3-Chloro-4-methyl)-3,3-dimethylurea, 1-(3-Chloro-4-methylphenyl)-3,3-dimethylurea, 3-(3-Chloro-4-methylphenyl)-1,1-dimethylurea, 3-(3-Chloro-p-tolyl)-1,1-dimethylurea, C 2242, CGA 15646, Chlorotoluron, Chlortoluron, Dicuran, Dicuran 500FL, Klortosan, Lentipur, Lentipur 500SC, Lentipur Flo, Lentipur Forte, N,N-Dimethyl-N'-(3-chloro-4-methylphenyl)urea, N-(3-Chloro-4-methylphenyl)-N',N'-dimethylurea, N-(3-Chloro-4-tolyl)-N',N'-dimethylurea, Shvat SC, Syncuran, Syncuran 80DP, Tolurex, Tolurgan
IUPAC Name 3-(3-chloro-4-methylphenyl)-1,1-dimethylurea
SMILES CN(C)C(=O)Nc1ccc(C)c(Cl)c1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C10H13ClN2O/c1-7-4-5-8(6-9(7)11)12-10(14)13(2)3/h4-6H,1-3H3,(H,12,14)
Formula Weight 212.0716
Chemical Name or Material Chlorotoluron
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.