missing translation for 'onlineSavingsMsg'
Learn More

Chlorothiazide, TRC

Product Code. 30134301
Change view
Click to view available options
Quantity:
50 mg
250 mg
2.5 g
Unit Size:
2.5g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134301 2.5 g 2.5g
17924585 250 mg 250mg
17924815 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134301 Supplier Toronto Research Chemicals Supplier No. TRCC3800002.5G
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-94-6
Molecular Formula C7 H6 Cl N3 O4 S2
Molecular Weight (g/mol) 295.72
Synonym 6-Chloro-2H-1,2,4-benzothiadiazine-7-sulfonamide 1,1-dioxide, 6-Chloro-7-sulfamoyl-2H-1,2,4-benzothiadiazine 1,1-dioxide, 6-Chloro-7-sulfamyl-1,2,4-benzothiadiazine 1,1-dioxide, Alurene, Chlorosal, Chlorothiazid, Chlorothiazide, Chlorthiazide, Chlorurit, Chlotride, Clotride, Diuresal, Diuril, Diuril Boluses, Diurilix, Diurite, Diutrid, Flumen, Minzil, NSC 25693, Neo-Dema, Salisan, Salunil, Saluretil, Saluric, Thiazide, Urinex, Warduzide, Yadalan
IUPAC Name 6-chloro-1,1-dioxo-2H-1λ^{6},2,4-benzothiadiazine-7-sulfonamide
SMILES NS(=O)(=O)c1cc2c(cc1Cl)N=CNS2(=O)=O

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C7H6ClN3O4S2/c8-4-1-5-7(2-6(4)16(9,12)13)17(14,15)11-3-10-5/h1-3H,(H,10,11)(H2,9,12,13)
Formula Weight 294.9488
Percent Purity >95%
Chemical Name or Material Chlorothiazide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.