missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Chlorosulfonyl)benzoic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | (Chlorosulfonyl)benzoic Acid |
| CAS | 10130-89-9 |
| Molecular Formula | C7H5ClO4S |
| Quantity | 5 g |
| Synonym | p-(Chlorosulfonyl)benzoic Acid, 4-(Chlorosulfonyl)benzoic Acid, 4-Carboxybenzenesulfonyl Chloride, 4-Carboxyphenylsulfonyl Chloride, NSC 191087, p-(Chlorosulfonyl)benzoic Acid, p-Carboxybenzenesulfonyl Chloride |
| InChI Formula | InChI=1S/C7H5ClO4S/c8-13(11,12)6-3-1-5(2-4-6)7(9)10/h1-4H,(H,9,10) |
| SMILES | OC(=O)c1ccc(cc1)S(=O)(=O)Cl |
| IUPAC Name | 4-chlorosulfonylbenzoic acid |
| Molecular Weight (g/mol) | 220.63 |
| Formula Weight | 219.9597 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?