missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide Solution 0.1M (0.1N), NIST Standard Solution ready to use, for volumetric analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™

Product Code. 10141860 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
10 L
Packaging:
Bag in box
HDPE plastic bottle
Unit Size:
1 L
10 L
2.5 L
5 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10141860 1 L HDPE plastic bottle 1 L
10558050 2.5 L HDPE plastic bottle 2.5 L
10637032 5 L HDPE plastic bottle 5 L
10224630 10 L Bag in box 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10141860 Supplier Fisher Chemical Supplier No. J/7660/15
Spotlight Offers
Use promo code 28036
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
This item is not returnable. View return policy

HNaO, CAS Number-1310-73-2, aetznatron, ascarite, caustic soda, soda, caustic, soda lye, sodium hydrate, sodium hydroxide, sodium hydroxide na oh, sodium hydroxide solution, white caustic, 1L, 100 deg.C, CHEBI:32145, Colorless, HEMHJVSKTPXQMS-UHFFFAOYSA-M, sodium;hydroxide, 3548, 39.997 | CAS: 1310-73-2 | HNaO | 39.997 g/mol

HNaO, CAS Number-1310-73-2, aetznatron, ascarite, caustic soda, soda, caustic, soda lye, sodium hydrate, sodium hydroxide, sodium hydroxide na oh, sodium hydroxide solution, white caustic, 1L, 100 deg.C, CHEBI:32145, Colorless, HEMHJVSKTPXQMS-UHFFFAOYSA-M, sodium;hydroxide, 3548, 39.997

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number 3548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Color Colorless
Boiling Point 100°C
Melting Point 0°C
Quantity 1 L
Packaging HDPE plastic bottle
Vapor Density > 1.0
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.