missing translation for 'onlineSavingsMsg'
Learn More

Chlorosulfonic acid, 97%

Product Code. 10397853 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
100mL
2.5 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10397853 2.5 L Glass bottle 2.5 L
10319170 100 mL Glass bottle 100mL
10512851 500 mL Glass bottle 500mL
10053740 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10397853 Supplier Thermo Scientific Acros Supplier No. 304490025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 7790-94-5 | ClHO3S | 116.515 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-94-5
InChI Key XTHPWXDJESJLNJ-UHFFFAOYSA-N
Synonym chlorosulfonic acid, chlorosulfuric acid, sulfuric chlorohydrin, chlorosulphuric acid, chlorosulphonic acid, monochlorosulfuric acid, unii-2o9axl1tj4, sulfonic acid, monochloride, chloridosulfuric acid, hsdb 909
PubChem CID 24638
IUPAC Name sulfurochloridic acid
SMILES OS(=O)(=O)Cl

Specifications

Density 1.7500g/mL
Packaging Glass bottle
Assay Percent Range 97%
Quantity 2.5 L
Molecular Formula ClHO3S
Linear Formula ClSO3H
Physical Form Oily Liquid, Fuming In Air
UN Number 1754
Fieser 01,140; 02,70; 06,121
Merck Index 15, 2168
Molecular Weight (g/mol) 116.515
Formula Weight 116.52
Percent Purity ≥96.0%
Chemical Name or Material Chlorosulfonic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Chlorosulfonic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Chlorosulfonic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.