Learn More
Palintest™ ChloroSense Chlorine Meter
Product Code. 11998109
Click to view available options
Range:
0.02 to 100ppm
Unit Size:
Each
Product Code. 11998109
Brand: Palintest™ CS100
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Simple, portable kit and instrument for acurate measurements of free and total chlorine in the field. Palintest™ ChloroSense Chlorine Meter is rugged and waterproof and can store 500 tests for future download.
  • Rugged, waterproof, field analysis instrument
  • Unique sensor technology for measuring free and total chlorine
  • Simple and rapid
  • No reagents or glassware required
  • Waterproof USB connection for instrument settings, software update and data download
  • Stores up to 500 test results
Specifications
For Use With (Application) Measure free and total chlorine
Includes USB port
Range 0.02 to 100ppm
Type ChloroSense Chlorine Meter