missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Chloroquine diphosphate salt, 98%

Product Code. p-7077537 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 15368865

Brand: Thermo Scientific Chemicals 455240250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chloroquine diphosphate salt, CAS # 50-63-5, is a member of the quinolone family that is considered to be a DNA intercalator. It shows anti-malarial, anti-inflammatory, and anti-filaricides activities. | CAS: 50-63-5 | C18H32ClN3O8P2 | 515.86 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Information


• Chloroquine diphosphate salt is a DNA intercalator that shows anti-malarial and anti-inflammatory activities.
• It has been described that this compound interferes with the biosynthesis of nucleic acids, suggesting a cellular mechanism of action


Applications

• Chloroquine diphosphate salt is used as a DNA intercalator to dissociate antigen-antibody complexes without denaturing red blood cell antigens
• This compound shows anti-malarial activity against Plasmodium falciparum, except against chloroquine-resistant strains. It has been used in in vitro antiplasmodial assays. In parasites the mechanism of action remains unclear, but this compound can inhibit the parasitic enzyme heme polymerase that converts the toxic heme into non-toxic hemozoin, resulting in the accumulation of toxic heme within the parasite.
• It shows very high anti-viral activity against NiV but lower activity against other viruses at concentrations lower than 20 μM
• Chloroquine diphosphate salt is used to study the role of endosomal acidification in cellular processes, such as the signaling of intracellular TLRs
• Chloroquine diphosphate salt has been used in autophagy inhibition studies

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-63-5
Molecular Formula C18H32ClN3O8P2
Molecular Weight (g/mol) 515.86
MDL Number MFCD00069852
InChI Key QKICWELGRMTQCR-UHFFFAOYNA-N
Synonym chloroquine diphosphate, chloroquine phosphate, aralen phosphate, chingamin phosphate, chloroquine bis phosphate, chingaminum, alermine, h-stadur, aralen diphosphate, miniquine
PubChem CID 64927
IUPAC Name 4-N-(7-chloroquinolin-4-yl)-1-N,1-N-diethylpentane-1,4-diamine;phosphoric acid
SMILES OP(O)(O)=O.OP(O)(O)=O.CCN(CC)CCCC(C)NC1=C2C=CC(Cl)=CC2=NC=C1

Specifications

Melting Point 200.0°C
Loss on Drying 2% max. (105°C, 16 h)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 25 g
Solubility Information Solubility in water: 50g/L. Other solubilities: insoluble in alcohol, benzene, chloroform, ether
Formula Weight 515.86
Percent Purity 98%
Chemical Name or Material Chloroquine diphosphate salt
compliance-icons
Product Identifier
  • Chloroquine diphosphate salt
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.