missing translation for 'onlineSavingsMsg'
Learn More

Chloropyramine hydrochloride

Product Code. 15493129
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15493129 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15493129 Supplier Thermo Scientific Alfa Aesar Supplier No. J60350.03
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6170-42-9 | C16H21Cl2N3 | 326.27 g/mol

Competitive reversible H-1 receptor antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 6170-42-9
Color White
Molecular Formula C16H21Cl2N3
MDL Number MFCD00079009
Quantity 1 g
Merck Index 14,2162
Synonym chloropyramine hydrochloride, chloropyramine hcl, alergosan, nilfan, allergan s hydrochloride, chlorpiramin hydrochloride, halopyramine hydrochloride, chlorpyramin hcl, chlorpyramine hydrochloride, unii-fwa92z14nn
InChI Key VEYWWAGBHABATA-UHFFFAOYSA-N
SMILES [H+].[Cl-].CN(C)CCN(CC1=CC=C(Cl)C=C1)C1=CC=CC=N1
IUPAC Name hydrogen N-[(4-chlorophenyl)methyl]-N-[2-(dimethylamino)ethyl]pyridin-2-amine chloride
Molecular Weight (g/mol) 326.27
PubChem CID 80311
Formula Weight 326.26
Physical Form Crystalline Powder
Chemical Name or Material Chloropyramine hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.