missing translation for 'onlineSavingsMsg'
Learn More

Chloroplatinic Acid Hexahydrate, ACS Reagent, ≥37.50% Pt basis, Solstice

Product Code. 15691770 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15691770 5 g Glass Bottle 5g
15671770 1 g Glass Bottle 1g
15681770 25 g Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15691770 Supplier Honeywell-Fluka Supplier No. 2060835G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ACS Reagent,≥37.50% Pt basis

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18497-13-7
Molecular Formula Cl6H14O6Pt
Molecular Weight (g/mol) 517.89
MDL Number MFCD00149910
InChI Key PIJUVEPNGATXOD-UHFFFAOYSA-J
IUPAC Name platinum(4+) dihydrogen hexahydrate hexachloride
SMILES [H+].[H+].O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Pt+4]

Specifications

Physical Form Powder
Quantity 5 g
Linear Formula H2PtCl6 · 6H2O
UN Number UN2507
Formula Weight 517.9g/mol
Percent Purity ≥37.50% (Pt basis)
Grade ACS Reagent
Packaging Glass Bottle
Chemical Name or Material Chloroplatinic Acid Hexahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.