missing translation for 'onlineSavingsMsg'
Learn More

Chloroplatinic Acid Hexahydrate ACS MP Biomedicals

Product Code. 11412812 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11412812 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11412812 Supplier MP Biomedicals Supplier No. 0215250905
  • ONLINE EXCLUSIVE PRICE
    £714.00
    £668.85 / 5g
    Save £45.15
    6% Off
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

CAS 18497-13-7
Quantity 5 g
Molecular Formula Cl6H14O6Pt
MDL Number MFCD00149910
Synonym Dihydrogen hexachloroplatinate
InChI Key PIJUVEPNGATXOD-UHFFFAOYSA-J
SMILES [H+].[H+].O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Pt+4]
IUPAC Name platinum(4+) dihydrogen hexahydrate hexachloride
Molecular Weight (g/mol) 517.89
Formula Weight 195.09
Grade ACS
Density 2.430 g/mL
Chemical Name or Material Chloroplatinic Acid, Hexahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.