missing translation for 'onlineSavingsMsg'
Learn More

Chloroplatinic Acid, 6-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Product Code. 15548064 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass Ampoule
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15548064 1 g Glass Ampoule 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15548064 Supplier Avantor J.T.Baker Supplier No. 289003

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Specifications

CAS 18497-13-7
Packaging Glass Ampoule
Molecular Formula Cl6H14O6Pt
MDL Number MFCD00149910
Quantity 1 g
Specific Gravity 1L = 2.43kg
InChI Key PIJUVEPNGATXOD-UHFFFAOYSA-J
SMILES [H+].[H+].O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Pt+4]
IUPAC Name platinum(4+) dihydrogen hexahydrate hexachloride
Molecular Weight (g/mol) 517.89
Formula Weight 517.92
Grade BAKER ANALYZED™ ACS Reagent
Physical Form Crystals
Chemical Name or Material Chloroplatinic Acid, 6-Hydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.