missing translation for 'onlineSavingsMsg'
Learn More

Chlorophyllin sodium copper salt, MedChemExpress

Product Code. 16448506
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16448506 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16448506 Supplier Medchem Express Supplier No. HYB0911
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Sodium copper chlorophyllin A is a semi-synthetic mixture of sodium copper salts derived from chlorophyll, primarily used as a food additive and in alternative medicine.

Sodium copper chlorophyllin A is a semi-synthetic mixture of sodium copper salts derived from chlorophyll, primarily used as a food additive and in alternative medicine.

Specifications

Chemical Name or Material Chlorophyllin sodium copper salt
Physical Form Solid
CAS 11006-34-1
Color Black
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C34H31CuN4Na3O6
For Use With (Application) Cancer-programmed cell death
Solubility Information H2O : ≥ 66.66 mg/mL (92.05 mM) ∣DMSO : 1.2 mg/mL (1.66 mM; Need ultrasonic)
SMILES O=C(O[Na])CC1=C(C(C(O[Na])=O)=C(C)C2=CC(C(CC)=C3C)=[N]4C3=CC5=C(C=C)C(C)=C6[N-]75)[N-]2[Cu+2]47[N](C8=C6)=C1C(C8C)CCC(O[Na])=O
Molecular Weight (g/mol) 724.15
Formula Weight 724.15
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.