missing translation for 'onlineSavingsMsg'
Learn More

Chlorophyllin, coppered trisodium salt

Product Code. 11403217
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11403217

Brand: Thermo Scientific Alfa Aesar A16155.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 11006-34-1 | C34H31CuN4Na3O6

Chlorophyllin, coppered trisodium salt is used in the spectrophotometric determination of Ven- lafaxine hydrochloride. A hybrid photoelectrochemical biofuel cell is composed of a TiO 2 film conductive glass electrode sensitized by a copper chlorophyll trisodium salt. Visible light-sensitive gels were prepared by introducing a chromophore (trisodium salt of copper chlorophyllin) to thermosensitive gels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorophyllin, coppered trisodium salt is used in the spectrophotometric determination of Ven- lafaxine hydrochloride. A hybrid photoelectrochemical biofuel cell is composed of a TiO 2 film conductive glass electrode sensitized by a copper chlorophyll trisodium salt. Visible light-sensitive gels were prepared by introducing a chromophore (trisodium salt of copper chlorophyllin) to thermosensitive gels.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11006-34-1
Molecular Formula C34H31CuN4Na3O6
MDL Number MFCD00012149

Specifications

Odor Odorless
Quantity 100 g
Solubility Information Soluble in water.
Formula Weight 724.15
Chemical Name or Material Chlorophyllin, coppered trisodium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.