missing translation for 'onlineSavingsMsg'
Learn More

Chlorophyll a (Technical Grade), TRC

Product Code. 30009961
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009961 10 mg 10mg
30009188 1 mg 1mg
30004377 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009961 Supplier Toronto Research Chemicals Supplier No. TRCC37988010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 479-61-8
Molecular Formula C55 H72 Mg N4 O5
Molecular Weight (g/mol) 893.49
Synonym Phorbine Magnesium Deriv., Chlorophyll a, Chlorophyll-1, Magnesium Pheophytin, Vegetable Chlorophyll, α-Chlorophyll, (SP-4-2)- [(2E,7R,11R)-3,7,11,15-tetramethyl-2-hexadecenyl (3S,4S,21R)-9-ethenyl-14-ethyl-21-(methoxycarbonyl)-4,8,13,18-tetramethyl-20-oxo-3-phorbinepropanoato(2-)-κN23,κN24,κN25,κN26]-Magnesium
IUPAC Name magnesium;methyl (3R,21S,22S)-16-ethenyl-11-ethyl-12,17,21,26-tetramethyl-4-oxo-22-[3-oxo-3-[(E,7R,11R)-3,7,11,15-tetramethylhexadec-2-enoxy]propyl]-23,25-diaza-7,24-diazanidahexacyclo[18.2.1.15,8.110,13.115,18.02,6]hexacosa-1,5,8(26),9,11,13(25),14,16,18,20(23)-decaene-3-carboxylate
SMILES CCC1=C(C)C2=N/C/1=C\c3c(C)c4C(=O)[C@H](C(=O)OC)\C\5=C/6\N=C(\C=C\7/N([Mg]n3c45)\C(=C/2)\C(=C7C)C=C)[C@@H](C)[C@@H]6CCC(=O)OC\C=C(/C)\CCC[C@H](C)CCC[C@H](C)CCCC(C)C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C55H73N4O5.Mg/c1-13-39-35(8)42-28-44-37(10)41(24-25-48(60)64-27-26-34(7)23-17-22-33(6)21-16-20-32(5)19-15-18-31(3)4)52(58-44)50-51(55(62)63-12)54(61)49-38(11)45(59-53(49)50)30-47-40(14-2)36(9)43(57-47)29-46(39)56-42;/h13,26,28-33,37,41,51H,1,14-25,27H2,2-12H3,(H-,56,57,58,59,61);/q-1;+2/p-1/b34-26+;/t32-,33-,37+,41+,51-;/m1./s1
Formula Weight 892.5353
Chemical Name or Material Chlorophyll-A
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.