missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Chlorophenol Red sodium salt

Product Code. 15498919
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15498919 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15498919 Supplier Thermo Scientific Chemicals Supplier No. L10364.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 123333-64-2 | C19H11Cl2O5S | 422.25 g/mol

Chlorophenol Red sodium salt is used in the standard samples to indicate pH. A stain used to detect chlorine dioxide in drinking water.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorophenol Red sodium salt is used in the standard samples to indicate pH. A stain used to detect chlorine dioxide in drinking water.

Solubility
Soluble in water.

Notes
Transition interval: pH 4.8
TRUSTED_SUSTAINABILITY

Specifications

CAS 123333-64-2
Melting Point ∽267°C (decomposition)
Molecular Formula C19H11Cl2O5S
MDL Number MFCD00151199
Quantity 5 g
Synonym chlorphenol red sodium salt
Solubility Information Soluble in water.
InChI Key QXTPRQZMDKBTAI-UNOMPAQXSA-M
SMILES OC1=CC=C(C=C1Cl)C(=C1/C=CC(=O)C(Cl)=C1)\C1=CC=CC=C1S([O-])(=O)=O
IUPAC Name 2-chloro-4-[3-(3-chloro-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol;sodium
Molecular Weight (g/mol) 422.25
PubChem CID 131846007
Formula Weight 445.26
Chemical Name or Material Chlorophenol Red sodium salt
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.