missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Chlorophenol Red, pure, indicator grade

Product Code. 10550804 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10550804 50 g Glass bottle 50g
10487580 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10550804 Supplier Thermo Scientific Chemicals Supplier No. 190040500
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4430-20-0 | C19H12Cl2O5S | 423.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4430-20-0
Molecular Formula C19H12Cl2O5S
Molecular Weight (g/mol) 423.26
MDL Number MFCD00005877
InChI Key WWAABJGNHFGXSJ-UHFFFAOYSA-N
Synonym CPR, 3', 3''-Dichlorophenylsulfonphthalein
PubChem CID 20486
IUPAC Name 2-chloro-4-[3-(3-chloro-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
SMILES OC1=CC=C(C=C1Cl)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(Cl)=C1

Specifications

Absorbance 1000 (at lambda max) (calc. on dried substance) (A (1% min./1cm))
Infrared Spectrum Authentic
Lambda Max 572 to 576nm (in 0.1 N NaOH)
Loss on Drying 8% max. (110°C)
Packaging Glass bottle
Specific Extinction 1000 min. (575nm, in 0.1N NaOH)
Visual Transition Interval From pH 4.8 (yellow) to pH 6.4 (purple)
Beilstein 19, II, 104
Color White to Cream
Solubility Information Solubility in water: insoluble
Quantity 50 g
Physical Form Granular Powder
Formula Weight 423.26
Grade Pure
Chemical Name or Material Chlorophenol Red, Pure
Show More Show Less
compliance-icons
Product Identifier
  • Chlorophenol Red
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.