missing translation for 'onlineSavingsMsg'
Learn More

Chloropentaamminecobalt(III) chloride

Product Code. 11351309
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11351309 50 g 50g
11331309 2 g 2g
11341309 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11351309 Supplier Thermo Scientific Alfa Aesar Supplier No. 013765.18
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13859-51-3

Chloropentaamminecobalt(III) chloride is doped with polyvinyl acetate to prepare polymeric films, which enhances the optical conductivity. It is also used to prepare the corresponding hydrogen sulfate complex by reacting with sulfuric acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13859-51-3
MDL Number MFCD00064728
Synonym chlorpentaaminecobalt, chloropentaamminecobalt iii dichloride, pentaaminechlorocobalt dichloride, pentaamminechlorocobalt dichloride, chloropentaminecobalt iii chloride, pentaaminechlorocobalt iii chloride, cobalt 3+ chloropentammine chloride, pentaamminechlorocobalt 2+ dichloride, cobalt 3+ , pentaammine-, trichloride, cobalt 2+ , pentaamminechloro-, dichloride, oc-6-22

Specifications

Melting Point (decomposition)
Color Purple
Density 1.819 g/mL
Odor Odorless
Linear Formula [CoCl(NH3)5]Cl2
Quantity 50 g
Solubility Information Soluble in water.
Formula Weight 250.44
Physical Form Powder
Chemical Name or Material Chloropentaamminecobalt(III) chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.