missing translation for 'onlineSavingsMsg'
Learn More

Chloromethyl Pivalate, TRC

Product Code. 30357731
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30357731 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30357731 Supplier Toronto Research Chemicals Supplier No. TRCC3696755G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chloromethyl Pivalate
CAS 18997-19-8
Molecular Formula C6H11ClO2
Quantity 5 g
Synonym 2,2-Dimethylpropanoic Acid Chloromethyl Ester, Chloromethanol Pivalate, (2,2-Dimethyl-1-oxopropoxy)methyl Chloride, 2,2-Dimethylpropionic Acid Chloromethyl Ester, Chloromethyl 2,2-dimethylpropanoate, Chloromethyl 2,2-Dimethylpropionate, Chloromethyl Pivalate, Chloromethyl Trimethylacetate, Pivaloyloxymethyl Chloride, [(tert-Butylcarbonyl)oxy]methyl Chloride, α,α-Dimethylpropionyloxymethyl Chloride
InChI Formula InChI=1S/C6H11ClO2/c1-6(2,3)5(8)9-4-7/h4H2,1-3H3
SMILES CC(C)(C)C(=O)OCCl
IUPAC Name chloromethyl 2,2-dimethylpropanoate
Molecular Weight (g/mol) 150.6
Formula Weight 150.0448
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.