missing translation for 'onlineSavingsMsg'
Learn More

Chloromethyl Ethyl Ether 90%, TRC

Product Code. 30350939
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30350939 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30350939 Supplier Toronto Research Chemicals Supplier No. TRCC369055100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chloromethyl Ethyl Ether
CAS 3188-13-4
Molecular Formula C3H7ClO
Quantity 100 g
Synonym Ethoxychloromethane, Ethoxymethyl Chloride, Ethyl Chloromethyl Ether, Chloromethoxyethane
InChI Formula InChI=1S/C3H7ClO/c1-2-5-3-4/h2-3H2,1H3
SMILES CCOCCl
IUPAC Name chloromethoxyethane
Molecular Weight (g/mol) 94.54
Formula Weight 94.0185
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.