missing translation for 'onlineSavingsMsg'
Learn More

Chloromethyl ethyl ether, 80%, Tech., stabilized

Product Code. 10294981 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10294981 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10294981 Supplier Thermo Scientific Acros Supplier No. 355720250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3188-13-4 | C3H7ClO | 94.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 3188-13-4
Color Colorless to Yellow
Density 1.0100g/mL
Boiling Point 82°C
Flash Point 19°C
Infrared Spectrum Authentic
Assay Percent Range 75% min. (GC)
Packaging Glass bottle
Molecular Formula C3H7ClO
Linear Formula CH3CH2OCH2Cl
Refractive Index 1.4000 to 1.405
MDL Number MFCD00000887
Quantity 25 mL
Specific Gravity 1.01
Synonym chloromethyl ethyl ether, chloromethoxy ethane, ethane, chloromethoxy, chloromethylethylether, ethoxymethyl chloride, ethyl chloromethyl ether, ethoxychloromethane, ether, chloromethyl ethyl, ethoxy methyl chloride
InChI Key FCYRSDMGOLYDHL-UHFFFAOYSA-N
SMILES CCOCCl
IUPAC Name chloromethoxyethane
Stabilizer CaCl2
Molecular Weight (g/mol) 94.54
PubChem CID 18523
Formula Weight 94.54
Percent Purity 80%
Grade Technical
Physical Form Liquid
Chemical Name or Material Chloromethyl ethyl ether, tech. 80%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Chloromethyl ethyl ether, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.