missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Chloromethyl)cyclopropane (Technical Grade), TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | (Chloromethyl)cyclopropane (Technical Grade) |
| CAS | 5911-08-0 |
| Molecular Formula | C4H7Cl |
| Quantity | 10 g |
| Synonym | 1-Chloromethylcyclopropane, Chlorocyclopropylmethane, Cyclopropanemethyl Chloride, Cyclopropylcarbinyl Chloride, Cyclopropylchloromethane, Cyclopropylmethyl Chloride, alpha-(Chloromethyl)cyclopropane |
| InChI Formula | InChI=1S/C4H7Cl/c5-3-4-1-2-4/h4H,1-3H2 |
| SMILES | ClCC1CC1 |
| IUPAC Name | chloromethylcyclopropane |
| Molecular Weight (g/mol) | 90.55 |
| Formula Weight | 90.0236 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction