missing translation for 'onlineSavingsMsg'
Learn More

Chloromethyl chlorosulfate, 97%

Product Code. 10652982 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Plastic bottle
Plastic bottle packed in tin can
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10652982 5 g Plastic bottle 5g
10325991 1 g Plastic bottle 1g
10003713 25 g Plastic bottle packed in tin can 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Product Code. 10652982 Supplier Thermo Scientific Acros Supplier No. 330800050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

CAS: 49715-04-0 | CH2Cl2O3S | 164.984 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 49715-04-0
MDL Number MFCD00041547
InChI Key PJBIHXWYDMFGCV-UHFFFAOYSA-N
Synonym chloromethyl chlorosulfate, chloromethyl chlorosulphate, chloromethylchlorosulfate, chloromethyl chlorosulfonate, chloromethyl chloranesulfonate, unii-n615783bnf, chloromethyl sulfochloridate, chlorosulfuric acid, chloromethyl ester, chloro chlorosulfonyloxy methane, chlorosulfuric acid chloromethyl ester
PubChem CID 2733556
IUPAC Name chloro(chlorosulfonyloxy)methane
SMILES C(OS(=O)(=O)Cl)Cl

Specifications

Color Colorless to Yellow
Density 1.6000g/mL
Boiling Point 45.0°C to 50.0°C (10.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Molecular Formula CH2Cl2O3S
Refractive Index 1.4460 to 1.4480
Quantity 5 g
Beilstein 01, III, 2593
Merck Index 15, 2147
Specific Gravity 1.6
Solubility Information Solubility in water: reacts.
Molecular Weight (g/mol) 164.99
Formula Weight 164.99
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Chloromethyl chlorosulfate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.