missing translation for 'onlineSavingsMsg'
Learn More

Chloromethyl butyrate, 99%

Product Code. 10175840 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10175840 25 g Glass bottle 25g
10643173 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10175840 Supplier Thermo Scientific Acros Supplier No. 295580250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 33657-49-7 | C5H9ClO2 | 136.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33657-49-7
Molecular Formula C5H9ClO2
Molecular Weight (g/mol) 136.58
MDL Number MFCD00216961
InChI Key BDPZFQLKFUONAG-UHFFFAOYSA-N
Synonym chloromethyl butyrate, chloromethylbutyrate, butyric acid chloromethyl ester, butanoic acid, chloromethyl ester, chloromethyl n-butanoate, butyryloxymethyl chloride, acmc-1agry, ksc492g7j, butanoic acid,chloromethyl ester, n-butanoic acid chloromethyl ester
PubChem CID 2725008
IUPAC Name chloromethyl butanoate
SMILES CCCC(=O)OCCl

Specifications

Color Colorless
Density 1.0740g/mL
Boiling Point 150.0°C (993.0 mbar)
Flash Point 55°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)2C(O)OCH2Cl
Refractive Index 1.4190 to 1.4210
Quantity 25 g
Specific Gravity 1.074
Solubility Information Solubility in water: 3g/L (20°C)
Formula Weight 136.58
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Chloromethyl butyrate, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Chloromethyl butyrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.