missing translation for 'onlineSavingsMsg'
Learn More

(Chloromethyl)benzene, Mikromol™

Product Code. 17949709
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17949709 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17949709 Supplier Mikromol Supplier No. MM0224.060500
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(Chloromethyl)benzene
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material alpha-Chlorotoluene (Benzylchloride)
CAS 100-44-7
Analyte or Component Names alpha-Chlorotoluene (Benzylchloride)
Shipping Condition Room Temperature
Molecular Formula C7 H7 Cl
Certifications/Compliance ISO 9001
Synonym Benzylchloride, Chloromethylbenzene, alpha-Chlorotoluene
InChI Formula InChI=1S/C7H7Cl/c8-6-7-4-2-1-3-5-7/h1-5H,6H2
SMILES ClCc1ccccc1
IUPAC Name chloromethylbenzene
Molecular Weight (g/mol) 126.58
Quantity 500 mg
Formula Weight 126.0236 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.